Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, Technical, 90%, Spectrum™

Click to view available options
Quantity:
500 mL
2.5 L
20 L
55 gal
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
64-18-6,7732-18-5
64-18-6,7732-18-5
CAS Max % 1
Percent Purity 0.9
Grade Technical
Packaging Poly Drum
Quantity 20 L

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.