Learn More
Formlabs IncSupplier Diversity Partner Clear Resin 1L  DFS Item
Manufacturer:  Formlabs IncSupplier Diversity Partner GLC02
Clear Resin polishes to near optical transparency, making it ideal for working with light or showcasing internal features. Supports print resolutions: 100, 50, 25 microns
Catalog No. NC1192437