Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formoterol Fumarate, Dihydrate, EP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
50 g
Packaging:
Amber Glass Bottle

F3015, 183814-30-4, (C19H24N2O4)2•C4H4O4•2H2O

Spectrum™ Formoterol Fumarate, Dihydrate, EP is a long-acting bronchodilator used as a long-term (maintenance) asthma treatment. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Optical Rotation -0.10° to +0.10°
Quantity 50 g
Water 4.0-5%
CAS 183814-30-4
CAS Max % 1
Percent Purity 98.5 to 101.5%