Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Formyl-Histone H1.3 (Lys140) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5121409

This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA
Formyl-Histone H1.3 (Lys140) Polyclonal antibody specifically detects Formyl-Histone H1.3 (Lys140) in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
Formyl-Histone H1.3 (Lys140) | |
Polyclonal | |
Unconjugated | |
H1-3 | |
H1 histone family, member 3; H1 VAR.4; H1.3; H1D; H1f3; H1s-2; H1s-4; Hist1h1d; histone 1, family 3; histone 1, H1d; histone cluster 1, H1d; histone H1.3; Histone H1c; histone H1s-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3007 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P16402 | |
H1-3 | |
Synthesized peptide derived from Human Histone H1.3 protein (137-149aa) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title