Alfa Aesar™ (Formylmethylene)triphenylphosphorane, 97%, may cont. up to »3% water

Manufacturer: Alfa Aesar™ A1170922

Quantity 100g

 View more versions of this product

Catalog No. AAA1170922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2136-75-6
    Molecular Formula (C6H5)3P=CHCHO
    Formula Weight 304.33
    MDL Number MFCD00006994
    Beilstein 523797
    Synonym (Triphenylphosphoranylidene)acetaldehyde
    Chemical Name or Material (Formylmethylene)triphenylphosphorane

    Specifications

    Formula Weight 304.33
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 2136-75-6
    Synonym (Triphenylphosphoranylidene)acetaldehyde
    Melting Point 183° to 188°C
    MDL Number MFCD00006994
    Beilstein 523797
    Health Hazard 1 Warning
    Recommended Storage 2°C to 8°C
    Concentration May contain up to ≈3% water
    Quantity 100g
    Molecular Formula (C6H5)3P=CHCHO
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material (Formylmethylene)triphenylphosphorane
    Assay 97%

    Description

    Applications
    (Formylmethylene)triphenylphosphorane is used as Wittig reagent. It is also used as Pharmaceutical Intermediates. It is employed as raw material in organic Synthesis, Agrochemicals.

    Solubility
    Soluble in Chloroform, Methanol.

    Notes
    Air Sensitive. Store at -20°C. Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit