Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Forskolin (From Coleus Forskohlii), Fisher BioReagents
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Packaging:
Amber Glass
Description
Forskolin is a molecular biology reagent. Forskolin functions as an antihypertensive and vasodilator. Adenylcyclase activator.
Specifications
Melting Point | 228°C |
Color | White |
Packaging | Amber Glass |
Quantity | 1 mg |
Merck Index | 15, 2460 |
Formula Weight | 410.5 |
Physical Form | Solid |
Chemical Name or Material | Forskolin, From Coleus forskohlii |
Safety and Handling
Recommended Storage | RT, desiccate |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction