Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Forsterite, Alfa Aesar™

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals

Manufacturer:  Alfa Aesar 4263903

Quantity 1g

 View more versions of this product

Catalog No. AA4263903


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Naturally Occurring Mineral, Grains, Approximately 0.06 To 0.19 in.
140.71
Forsterite
Mg2SiO4
MFCD00283278

Specifications

Naturally Occurring Mineral, Grains, Approximately 0.06 To 0.19 in.
140.71
MFCD00283278
Light green
Forsterite
Mg2SiO4
Light green chunks
Odorless
1g

Safety and Handling

SDS & Safety Documents

H332: Harmful if inhaled.

EINECSNumber : 215-681-1

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit