Forsterite, Alfa Aesar™

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals

Manufacturer: Alfa Aesar 4263906

Quantity 5g

 View more versions of this product

Catalog No. AA4263906

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Naturally Occurring Mineral, Grains, Approximately 0.06 To 0.19 in.
    Molecular Formula Mg2SiO4
    Formula Weight 140.71
    MDL Number MFCD00283278
    Chemical Name or Material Forsterite

    Specifications

    Physical Form Naturally Occurring Mineral, Grains, Approximately 0.06 To 0.19 in.
    Formula Weight 140.71
    Quantity 5g
    Color Light green
    Density 3.21g/cm3
    EINECS Number 215-681-1
    Odor Odorless
    Molecular Formula Mg2SiO4
    Appearance Light green chunks
    MDL Number MFCD00283278
    Health Hazard 1 H332: Harmful if inhaled.
    Chemical Name or Material Forsterite

    H332: Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit