Learn More
FOSB Mouse anti-Human, Clone: OTI2A1, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF804112
Catalog No. 50-166-6815
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
FOSB
Monoclonal
Unconjugated
FOSB
Lyophilized
FOSB
Human recombit protein fragment corresponding to amino acids 1-255 of human FOSB produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI2A1
PBS with 8% trehalose and no preservative; pH 7.3
P53539
AP-1, G0S3, GOS3, GOSB
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2354