Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FosB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA594997
Description
Synthetic peptide sequence: 1-15aa, MFQAFPGDYDSGSRC. Reconstitution information: Add 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
c-FOS is a nuclear phosphoprotein which forms a tight but non-covalently linked complex with the JUN/AP-1 transcription factor. It has a critical function in regulating the development of cells destined to form and maintain the skeleton, and is thought to have an important role in signal transduction, cell proliferation and differentiation.Specifications
FosB | |
Polyclonal | |
0.2mg sodium phosphate with 0.9MG NaCl, 0.9MG NaCl, 5MG BSA, 5MG BSA and 0.05MG sodium azide, 0.05MG sodium azide, 0.05MG thimerosal, 0.05MG thimerosal | |
P13346, P53539 | |
FOSB | |
A synthetic peptide corresponding to a sequence at the N-terminus of human Fos B, identical to the related mouse and rat sequences. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
FOSB | |
Protein fosB; G0/G1 switch regulatory protein 3; FOSB; G0S3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100360880, 14282, 2354 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title