Learn More
FOSL2 Mouse anti-Human, Clone: OTI1C6, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809663
Catalog No. 50-166-9639
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation.
Specifications
Specifications
FOSL2
Monoclonal
Unconjugated
FOSL2
Lyophilized
FOSL2
Human recombit protein fragment corresponding to amino acids 110-326 of human FOSL2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1C6
PBS with 8% trehalose and no preservative; pH 7.3
P15408
FRA2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2355