Learn More
Foss Exhaust Manifold for Digestors
Facilitate fume removal and containment
$2432.00 - $2955.00
Specifications
For Use With (Application) | For use with all digestion procedures |
---|---|
For Use With (Equipment) | Digestion unit |
Description
Ensures Good Laboratory Practice (GLP).
- Strongly recommended for use with all digestion procedures
- Should be connected to water aspirator (supplied) or to a suitable scrubber unit
Specifications
For use with all digestion procedures | |
Digestion unit |