Learn More
Foss™ Digestion System: Scrubber Units: SR210
For neutralization of corrosive fumes in acid digestions
Manufacturer:  Foss™ 60063227
Catalog No. 12-053-110
Add to cart
Description
Description
  • Can be combined with any of the exhaust manifolds
  • Handles up to 100 samples without changing of reagent
  • Compact benchtop unit is self-contained and therefore unaffected by water supply issues
  • Manuall controlled adjustable setting
Specifications
Specifications
50/60 Hz
230 V, 50/60 Hz
Acid Digester
230 V
Semi-automated neutralization of corrosive fumes in acid digestions
1 Ea.