Learn More
Foss™ Soxtec™ 8000 Extraction Systems for Extractable Matter
For fast and safe extraction of soluble matter from wide range of matrices
Includes: Accessories Include: 3 sets glass extraction cups 6/set, 1 set condenser seals, 2 thimble stands, 2 set thimble adapters 6/set, docking tool for thimbles, set of cellulose thimbles 33mm double thickness 25/set, thimble support, cup holder, cup stand, cup tool,cotton, solvent addition tube, solvent recovery flask, owners guide, application note, user manual, quick guide, spare parts manual
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
TCS8000800G
View Documents
Foss™
S8000800G
6 position, 1 basic unit, 1 control unit, 1 set of accessories 6 position, 1 basic unit, 1 control unit, 1 set of accessories This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Fully automatic for unattended operation
  • Consists of basic unit and control unit, glass cups included
  • 6 hot plate positions, expandable up to 12
  • Sample size: 0.5 to 15g, range is application dependent
  • 100 to 120VAC or 200 to 240VAC, depending on version, 50/60 Hz
  • Power: 1500w. (120VAC version), 1800w. (230VAC version)
  • D x W x H: 350 x 640 x 630mm (extraction unit)
  • Closed solvent addition limits operator's exposure to solvent
  • Extraction cups use 70-90mL solvent, 80% of which is recovered
  • No touching or mixing of samples is ensured with the batch handling tools
  • Cooling water control reduces water costs
  • Flexible operation
  • Safe and consistent solvent dosage
  • Consistent solvent volume
Certifications
CE marked