Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
American Educational Products Fossil Identification Lab
Uncover actual fossils more than 500 million years old.
Specifications
For Use With (Application) | Uncover actual fossils more than 500 million years old |
---|---|
For Use With (Equipment) | 15 Students |
Grade | 5 to 12 |
Includes: Excavation tools, fossil card deck, teacher's guide and a student reference guide
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity & Availability | ||||
S53501
![]() |
American Educational Products
3770B |
Fossil Identification | 15 Replacement fossil/rocks with 4 fossils each In a sealable Plastic bag |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
Description
- Use the same scientific processes as skilled geologists
- Dig through rocks (made of plaster) to find genuine fossil fragments
Order Info
Class size: 15 students.
Specifications
Uncover actual fossils more than 500 million years old | |
15 Students | |
5 to 12 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title