Learn More
Selleck Chemical LLC Fostamatinib (R788) S2625-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S262510MM/1ML
Encompass_Preferred
Fostamatinib (R788) a prodrug of the active metabolite R406 is a Syk inhibitor with IC50 of 41 nM strongly inhibits Syk but not Lyn 5-fold less potent to Flt3 Phase 3
Catalog No. 50-311-2509