Learn More
Fotodyne™ FOTO/Analyst™ Luminary/FX Workstations with Single Wavelength UV Transilluminator
Transillumination and epi-illumination with a variety of emission options and large format transilluminators give you flexibility to capture images of many different lab samples
Manufacturer:  Fotodyne™ 71720DLNP
 View more versions of this product
Catalog No. F71720DLN
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CCD camera combined with light-tight FOTO/FX Benchtop Darkroom and extended integration software allow you to maximize exposure times for low-light chemilumiescent samples
  • Acquire and store digital images of chemiluminescent, fluorescent, and visible samples, including gels, western blots, TLC plates, and Petri dishes
  • Hi-QE camera delivers 60-70% quantum efficiency (QE) at 425-625nm; Firewire connection provides fast downloads
  • Offer picogram-level sensitivity for both nucleic acid and protein gels and blots
  • Raised platform brings samples closer to the imaging source, increasing sensitivity while limiting distortion
FOTO/FX™ Benchtop Darkroom
  • Houses a cooled CCD camera with a hands-free motorized zoom lens
  • Gliding transilluminator platform and side-opening door with interlock provide convenient and safe operation
  • Choose workstations with either UV or dual light transilluminators
  • Lumi/Tray is designed specifically for work with chemiluminescence and positions blots close to the camera for maximum sensitivity
  • Dual visible epi-lights allow easy focusing and even illumination of opaque samples
  • Motorized filter wheel with position for five filters
  • Optional dual-UV and RGB epi-lights
FOTO/Analyst PC Image Acquisition Software
  • Captures and stores images in a choice of nine file formats, including TIFF, JPEG, and BMP
  • Offers background subtraction, brightness and contrast adjustment, image inversion, and a saturation indicator to determine when images are exposed beyond the limits of effective digital analysis
  • Annotation feature permits labeling with text, date and time stamp, arrows, circles, and boxes
  • Up to five exposure times can be set to capture consecutive images of low-signal samples for optimal results
Minimum Computer Requirements
  • Windows™ XP™, Vista™, or 7; 2.0GHz or faster processor, 1GB RAM; 20GB hard drive space (additional space for images)
  • One open full-size PCI slot, two open USB ports; open serial port
  • 17 in. monitor
  • 1024 x 768 resolution video card, 24-bit true color
Order Info
Available with or without thermal printer. Standard size transilluminator measures 21 x 26cm.
Specifications
Specifications
2 Mpx
No
Standard size
Single-wavelength UV, dual light transilluminator