Learn More
GE Healthcare Four Gel Caster Available on GSA/VA Contract for Federal Government customers only.
Modular design of miniVE vertical electrophoresis system consolidating all small parts for electrophoresis and blotting into one unit.
Manufacturer:  GE Healthcare 80614612
Catalog No. 45-002-435
Add to cart
Description
Description
  • Uniform gel temperature maintained with tank-style lower buffer chamber
  • Lower buffer chamber holds two Blot Modules, allowing four gels to be blotted at once
  • Gel module acts as a casting stand and upper buffer chamber with integrated clamps
  • Handles gel cassettes up to 10W x 10.5cm H
  • Compatible with glass plates, combs, and spacers of SE 260 Mini-Vertical units
  • EPS 301 is preferred power supply for miniVE System
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Cast up to 4 gels
4
0.75 mm, 1.0 mm and 1.5 mm
4-Gel Caster Complete for 2–4 Gels, 10 × 10.5 cm Plates
4 x Notched alumina plate,10 x 10.5 cm, 5 x Rectangular glass plate, 10 x 10.5 cm, 4 x Clamp, 100 x Wax paper sheet, 1 x Space-saver plate, 5 x Filler sheet, 1 x Filler plugs set , 1 x Spacer-Mate assembly template
Complete packsize
Gel Cassettes up to 10 x 10.5 cm