Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXA1 Mouse anti-Human, Clone: FOXA1/1515, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14557

Catalog No. 89-034-640


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human FOXA1.

  • High specificity
  • High reactivity
Specifications

Specifications

FOXA1
FOXA1/1515
Mouse monoclonal antibody raised against partial recombinant human FOXA1.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
FOXA1
Recombinant protein corresponding to amino acids 467-614 of human FOXA1.
Protein A/G purification
RUO
Primary
3169
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
P55317
HNF3A|MGC33105|TCF3A
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)