Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXB1 Mouse anti-Human, Janelia Fluor 549, Clone: PCRP-FOXB1-1B7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314021JF549
This item is not returnable.
View return policy
Description
FOXB1 Monoclonal antibody specifically detects FOXB1 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
FOXB1 | |
Monoclonal | |
Janelia Fluor 549 | |
FKH5HFKH-5, forkhead box B1, forkhead box protein B1, Transcription factor FKH-5 | |
Recombinant full-length human FOXB1protein (Uniprot: Q99853) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-FOXB1-1B7 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
27023 | |
Store at 4°C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title