Learn More
FOXB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 242851AP
Catalog No. 50-173-4277
Add to cart
Description
Description
Foxb1 gene ontology annotations related to this gene include anatomical structure morphogenesis; axon target recognition; cell differentiation; cell migration in diencephalon; epithelial cell differentiation involved in mammary gland alveolus development; floor plate development; hypothalamus cell migration; inferior colliculus development; lactation; mammary gland lobule development; mammillary body development; mammillothalamic axonal tract development; midbrain development; negative regulation of neuron apoptotic process; regulation of transcription by RNA polymerase II; somitogenesis; spinal cord development; telencephalon cell migration; thalamus development; urogenital system development; visual learning.
Specifications
Specifications
FOXB1
0.51 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q64732, Q99853
FKH5, forkhead box B1, Forkhead box protein B1, FOXB1, HFKH 5, Transcription factor FKH 5
Rabbit
IgG
150 μL
-20°C
Primary
27023, 64290
Western Blot
Unconjugated
FOXB1
Liquid
Foxb1
FOXB1 Fusion Protein Ag18079
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse