Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxC2 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI3E3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270752AF700

Catalog No. N270752A700


Add to cart

Description

Description

FoxC2 Monoclonal antibody specifically detects FoxC2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

FoxC2
OTI3E3
Western Blot
Affinity Purified, Conjugated
FOXC2
Recombinant protein fragment corresponding to amino acids 1-280 of human FoxC2 (NP_005242) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
FKHL14LD, forkhead box C2 (MFH-1, mesenchyme forkhead 1), forkhead, Drosophila, homolog-like 14, Forkhead-related protein FKHL14, Mesenchyme fork head protein 1, mesenchyme forkhead 1, MFH-1, MFH-1 protein, MFH1forkhead box protein C2, Transcription factor FKH-14
Mouse
IgG1
0.1 mL
Primary
2303.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit