Learn More
FoxC2 Mouse anti-Human, DyLight 405, Clone: 1D11C8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237409V
Catalog No. NBP237409V
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
FoxC2
1D11C8
Western Blot, ELISA
Purified
FOXC2
Purified recombinant fragment of human FOXC2 (AA: 21-210) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
FKHL14LD, forkhead box C2 (MFH-1, mesenchyme forkhead 1), forkhead, Drosophila, homolog-like 14, Forkhead-related protein FKHL14, Mesenchyme fork head protein 1, mesenchyme forkhead 1, MFH-1, MFH-1 protein, MFH1forkhead box protein C2, Transcription factor FKH-14
Mouse
IgG1
0.1 ml
Primary
2303