Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXD1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550498

Catalog No. PA550498


Add to cart

Description

Description

The antibody detects endogenous levels of total FOXD1 protein.

This intronless gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in tumor formation.
Specifications

Specifications

FOXD1
Polyclonal
Unconjugated
FOXD1
Liquid
FOXD1
Synthetic peptide corresponding to residues near the N terminal of human Forkhead box protein D1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q16676, Q61345
FKHL8, FREAC4
Rabbit
IgG
100 μL
-20°C
Primary
15229, 2297
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit