Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXD1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP31040425UL

 View more versions of this product

Catalog No. NB125710


Add to cart

Description

Description

FOXD1 Polyclonal antibody specifically detects FOXD1 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

FOXD1
Unconjugated
PBS buffer, 2% sucrose
FKHL8Forkhead, drosophila, homolog-like 8, forkhead box D1, forkhead-like 8, forkhead-related activator 4, Forkhead-related protein FKHL8, Forkhead-related transcription factor 4, FREAC-4, FREAC4forkhead box protein D1
The immunogen is a synthetic peptide directed towards the N terminal region of mouse FOXD1 (NP_032268). Peptide sequence EGEDDEEEEDDDDEGGGGRGGGGSRLPSSAQRRRRSYAGEDDLEDLEEED
25 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Primary
2297
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit