Learn More
FoxF2 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF6988SP
Catalog No. AF6988SP
Add to cart
This item is not returnable. View return policy
Description
Description
FoxF2 Polyclonal antibody AF6988SP specifically detects FoxF2 in Human samples. It is validated for Western Blotting
Specifications
Specifications
FoxF2
LYOPH
Western Blot 1 ug/mL
Q12947
FKHL6, FKHL6forkhead box protein F2, forkhead box F2, forkhead-like 6, Forkhead-related activator 2, Forkhead-related protein FKHL6, Forkhead-related transcription factor 2, FREAC2, FREAC2FREAC-2
Sheep
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human FoxF2 in direct ELISAs and Western blots. In direct ELISAs, less than 3% cross-reactivity with recombinant human (rh) FoxF1, rhFoxP3, and rhFoxC2 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FOXF2
E. coli-derived recombinant human FoxF2 Ala324-Met444 Accession # Q12947
Immunogen affinity purified
RUO
Primary
2295
Sterile PBS to a final concentration of 0.2 mg/mL.