Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXG1 Polyclonal Antibody, Huabio
Rabbit Polyclonal Antibody
Supplier: HUABIO RESEARCH INC HA500064


Description
FOXG1 Polyclonal antibody specifically detects FOXG1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
FOXG1 | |
Polyclonal | |
Unconjugated | |
FOXG1 | |
2900064B05Rik; Bf1; BF-1; BF1A; BF2; BF-2; brain factor 1; Brain factor 2; FHKL3; FKH2; FKHL1; FKHL2; FKHL3; FKHL4; forkhead box G1; forkhead box G1B; forkhead box protein G1; Forkhead box protein G1A; Forkhead box protein G1B; Forkhead box protein G1C; forkhead-like 1; forkhead-like 2; forkhead-like 3; forkhead-like 4; forkhead-like transcription factor BF-1; forkhead-related protein FKHL1; Forkhead-related protein FKHL2; Forkhead-related protein FKHL3; FOXG1; FOXG1A; FOXG1B; FOXG1C; HBF-1; HBF2; HBF-2; HBF-3; HBF-G2; Hfh9; Hfhbf1; HFK1; HFK2; HFK3; HNF-3/forkhead homolog, brain factor 1; KHL2; oncogene QIN; QIN; RATBF1A; transcription factor BF-1 | |
Synthetic peptide within human FOXG1 aa 430-485. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P55316, Q60987 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15228, 2290 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction