Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxH1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen FoxH1
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125704
View Documents
Novus Biologicals
NBP31040125UL
25 μg
Each for $106.19
Add to cart
 
NB125703
View Documents
Novus Biologicals
NBP310401100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

FoxH1 Polyclonal antibody specifically detects FoxH1 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

FoxH1
Western Blot
Purified
RUO
Mouse
Fast-1, FAST1forkhead box protein H1, Fast-2, FAST2, Forkhead activin signal transducer 1, Forkhead activin signal transducer 2, forkhead activin signal transducer-1, forkhead box H1, hFAST-1, TGF-beta/activin signal transducer
The immunogen is a synthetic peptide directed towards the middle region of mouse FoxH1 (NP_032015). Peptide sequence GPSSSSETPLWPLCSLPGPTIIEGESSQGEVIRPSPVTPDQGSWPLHLLE
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Stem Cell Markers
PBS buffer, 2% sucrose
8928
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit