Learn More
FoxH1 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF4248SP
 View more versions of this product
Catalog No. AF4248SP
Add to cart
This item is not returnable. View return policy
Description
Description
FoxH1 Polyclonal antibody AF4248SP Specifically detects FoxH1 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
FoxH1
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
O75593
FAST1, FAST-1, FAST1forkhead box protein H1, Fast-2, FAST2, Forkhead activin signal transducer 1, Forkhead activin signal transducer 2, forkhead activin signal transducer-1, forkhead box H1, hFAST-1, TGF-beta/activin signal transducer
Sheep
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human FoxH1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human FoxB2 is observed.
Human
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FOXH1
E. coli-derived recombinant human FoxH1 Ser164-Ser272 Accession # O75593
Immunogen affinity purified
RUO
Primary
8928
Reconstitute at 0.2 mg/mL in sterile PBS.