Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOXJ1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580643
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human FOXJ1.
Forkhead-box J1 (FOXJ1) is a 421-amino acid transcription factor that suppresses T cell activity and spontaneous autoimmunity, through the repression of NF B activity. FOXJ1 also inhibits the humoral immune response in B cells. FOXJ1 deficiency in B cells results in spontaneous and accentuated germinal center formation and is implicated in the development of pathogenic autoantibodies and accentuated responses to immunizations. Abnormal expression of FOXJ1 may be associated with autoimmune diseases and/or other inflammatory diseases. FOXJ1 is also required for cilia formation and left-right axis determination because it increases calpastatin expression, a protein necessary for the ability of basal bodies to anchor to the apical cytoskeleton. FOXJ1 expression may function as an early marker of epithelial cell differentiation, recovery, and function.
Specifications
FOXJ1 | |
Polyclonal | |
Unconjugated | |
FOXJ1 | |
FKHL13; FKHL-13; fork head homologue 4; forkhead box J1; forkhead box protein J1; forkhead transcription factor HFH-4; forkhead-like 13; forkhead-related protein FKHL13; Foxj1; Hepatocyte nuclear factor 3 forkhead homolog 4; Hfh4; HFH-4; HNF-3/forkhead homolog 4; MGC35202 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
2302 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q92949 | |
FOXJ1 | |
A synthetic peptide corresponding to the center region of the Human FOXJ1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction