Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXJ3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548557
Description
Predicted to react with mouse based on sequence homology.
Transcriptional activator of MEF2C involved in the regulation pf adult muscle fiber type identity and skeletal muscle regeneration.
Specifications
FOXJ3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9UPW0 | |
FOXJ3 | |
KLH conjugated synthetic peptide between 148-176 amino acids from the N-terminal region of human FOXJ3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
FOXJ3 | |
Forkhead box protein J3, FOXJ3, KIAA1041 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22887 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction