Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXL2 Rabbit anti-Human, PE/Atto594, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP270013PEATT594

Catalog No. N27013PE594


Add to cart

Description

Description

FOXL2 Polyclonal antibody specifically detects FOXL2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

FOXL2
PE/ATTO 594
PBS with 0.05% Sodium Azide
BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3
Rabbit
IgG
0.1 mL
Store at 4°C in the dark. Do not freeze.
Polyclonal
Human
Flow Cytometry
Flow Cytometry
Affinity Purified, Conjugated
FOXL2
Synthetic peptide made to a C-terminal portion of the human FOXL2 protein (between amino acids 350-376) [UniProt P58012]
Immunogen affinity purified
Epigenetics
Primary
668.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit