Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXL2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen FOXL2
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095697
View Documents
Novus Biologicals
NBP303941100UL
100 μg Each for $320.71
Add to cart
 
NB095698
View Documents
Novus Biologicals
NBP30394120UL
20 μg Each for $138.37
Add to cart
 
Description

Description

FOXL2 Polyclonal antibody specifically detects FOXL2 in Human samples. It is validated for Western Blot
Specifications

Specifications

FOXL2
Unconjugated
Rabbit
Epigenetics
PBS with 50% glycerol, pH7.3.
668
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human
BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3
A synthetic peptide corresponding to a sequence within amino acids 300 to the C-terminus of human FOXL2 (NP_075555.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit