Learn More
Aviva Systems Biology Corp FOXO1 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp P100607_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XO1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FOXO1. Alternate names: FKH1, FKHR, FOXO1AApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-62660
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.