Learn More
FoxO1/FKHR Mouse anti-Human, Janelia Fluor 549, Clone: 7H11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141197JF549
Catalog No. NBP14197549
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
FoxO1/FKHRMonoclonalantibody specifically detects FoxO1/FKHR in Human samples.
Specifications
Specifications
FoxO1/FKHR
7H11
Immunohistochemistry
Purified
FOXO1
Amino acids 242-655 of FKHR expressed in E. coli.
Protein G purified
Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers
Primary
2308
Human
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma
Mouse
IgM
0.1mL
Store at 4°C in the dark.
Monoclonal
This antibody will recognize human FKHR.