Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO1A/3/4 Mouse anti-Human, FITC, Clone: 40000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141196F

Catalog No. NBP141196F


Add to cart

Description

Description

FOXO1A/3/4 Monoclonal antibody specifically detects FOXO1A/3/4 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

FOXO1A/3/4
40000000
Immunohistochemistry
Affinity Purified
FOXO1
Amino acids 242-655 of FKHR expressed in E. coli.
Immunogen affinity purified
Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers
Primary
2308
Human
Immunohistochemistry
FITC
PBS with 0.05% Sodium Azide
FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
This antibody will recognize human FKHR.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit