Learn More
FOXO1A/3/4 Mouse anti-Human, Janelia Fluor 646, Clone: 40000000, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141196JF646
Catalog No. NBP14196646
Add to cart
Description
Description
FOXO1A/3/4 Monoclonal antibody specifically detects FOXO1A/3/4 in Human samples. It is validated for Immunohistochemistry.
Specifications
Specifications
FOXO1A/3/4
40000000
Immunohistochemistry
Purified
FOXO1
Amino acids 242-655 of FKHR expressed in E. coli.
Protein G purified
Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers
Primary
2308
Human
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
This antibody will recognize human FKHR.