Learn More
FOXO4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 215351AP
Catalog No. 50-173-3469
Add to cart
Description
Description
This gene encodes a member of the O class of winged helix/forkhead transcription factor family. Proteins encoded by this class are regulated by factors involved in growth and differentiation indicating they play a role in these processes. A translocation involving this gene on chromosome X and the homolog of the Drosophila trithorax gene, encoding a DNA binding protein, located on chromosome 11 is associated with leukemia. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
FOXO4
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P98177, Q9WVH3
AFX, AFX1, forkhead box O4, Forkhead box protein O4, FOXO4, MLLT7
Rabbit
IgG
150 μL
-20°C
Primary
302415, 4303, 54601
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FOXO4
Liquid
Foxo4
FOXO4 Fusion Protein Ag16169
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat