Learn More
FOXP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA551087
Catalog No. PA551087
Add to cart
Description
Description
FOXP1 Polyclonal Antibody detects endogenous levels of FOXP1 protein.
Transcriptional repressor (PubMed:18347093, PubMed:26647308). Can act with CTBP1 to synergistically repress transcription but CTPBP1 is not essential. Plays an important role in the specification and differentiation of lung epithelium. Acts cooperatively with FOXP4 to regulate lung secretory epithelial cell fate and regeneration by restricting the goblet cell lineage program; the function may involve regulation of AGR2. Essential transcriptional regulator of B-cell development. Involved in regulation of cardiac muscle cell proliferation. Involved in the columnar organization of spinal motor neurons. Promotes the formation of the lateral motor neuron column (LMC) and the preganglionic motor column (PGC) and is required for respective appropriate motor axon projections. The segment-appropriate generation of spinal chord motor columns requires cooperation with other Hox proteins. Can regulate PITX3 promoter activity; may promote midbrain identity in embryonic stem cell-derived dopamine neurons by regulating PITX3. Negatively regulates the differentiation of T follicular helper cells T(FH)s. Involved in maintaice of hair follicle stem cell quiescence; the function probably involves regulation of FGF18. Represses transcription of various pro-apoptotic genes and cooperates with NF-kappa B-signaling in promoting B-cell expansion by inhibition of caspase-dependent apoptosis (PubMed:25267198). Binds to CSF1R promoter elements and is involved in regulation of monocyte differentiation and macrophage functions; repression of CSF1R in monocytes seems to involve NCOR2 as corepressor (PubMed:15286807, PubMed:18799727, PubMed:18347093). Involved in endothelial cell proliferation, tube formation and migration indicative for a role in angiogenesis; the role in neovascularization seems to implicate suppression of SEMA5B (PubMed:24023716). Can negatively regulate androgen receptor signaling (PubMed:18640093). {ECO:0000250; UniProtKB:P58462, ECO:0000269; PubMed:15286807, ECO:0000269; PubMed:18640093, ECO:0000269; PubMed:18799727, ECO:0000269; PubMed:24023716, ECO:0000269; PubMed:25267198, ECO:0000269; PubMed:26647308, ECO:0000305; PubMed:18347093, ECO:0000305; PubMed:24023716}.; Isoform 8: Involved in transcriptional regulation in embryonic stem cells (ESCs). Stimulates expression of transcription factors that are required for pluripotency and decreases expression of differentiation-associated genes. Has distinct DNA-binding specifities as compared to the canonical form and preferentially binds DNA with the sequence 5 ft.-CGATACAA-3 ft. (or closely related sequences) (PubMed:21924763). Promotes ESC self-renewal and pluripotency. {ECO:0000250; UniProtKB:P58462, ECO:0000269; PubMed:21924763}.
Specifications
Specifications
FOXP1
Polyclonal
Unconjugated
FOXP1
Liquid
FOXP1
Synthesized peptide derived from the C-terminal region of human FOXP1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9H334, P58462, Q498D1
12CC4, FLJ23741, HSPC215, MGC12942, MGC88572, MGC99551, QRF1, hFKH1B
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
108655, 27086, 297480