Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP1 Rabbit anti-Human, Clone: FOXP1/44R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen FoxP1
Clone FOXP1/44R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122068
View Documents
Novus Biologicals
NBP30729720UG
20 μg
Each for $159.82
Add to cart
 
NB122067
View Documents
Novus Biologicals
NBP30729710UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

FoxP1 Monoclonal antibody specifically detects FoxP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

FoxP1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
27086
IgG Kappa
Primary
Store at 4°C.
FOXP1/44R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Human
FLJ23741, fork head-related protein like B, forkhead box P1, forkhead box protein P1, glutamine-rich factor 1, hFKH1B, HSPC215, MGC12942, MGC88572, MGC99551, QRF112CC4
Synthetic peptide corresponding to residues within aa350-450 of FoxP1
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit