Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXQ1 Mouse anti-Human, DyLight 550, Clone: PCRP-FOXQ1-2D2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314070R
This item is not returnable.
View return policy
Description
FOXQ1 Monoclonal antibody specifically detects FOXQ1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
FOXQ1 | |
Monoclonal | |
DyLight 550 | |
forkhead box Q1, Hepatocyte nuclear factor 3 forkhead homolog 1, HFH-1, HFH1forkhead box protein Q1, HNF-3/forkhead-like protein 1, winged helix/forkhead transcription factor | |
Recombinant full-length human FOXQ1 protein (Uniprot: Q9C009) | |
0.1 mL | |
Cancer | |
94234 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-FOXQ1-2D2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title