Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXR1 Monoclonal Antibody (OTI5A5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF812053


Description
FOXR1 Monoclonal antibody specifically detects FOXR1 in Human samples. It is validated for Western BlotSpecifications
FOXR1 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P55822 | |
Mouse | |
Protein A/G | |
RUO | |
6450 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Western Blot | |
OTI5A5 | |
Unconjugated | |
Sh3bgr | |
DLNB13; Forkhead box protein N5; forkhead box protein R1; forkhead box R1; forkhead box R1 variant 1; forkhead box R1 variant 2; Foxn5; FOXR1; Gm1115 | |
Full length human recombinant protein of human SH3BGR (NP_031367) produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction