Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXR2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | FOXR2 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180511
|
Novus Biologicals
NBP180511 |
100 μL |
Each of 1 for $436.00
|
|
Description
FOXR2 Polyclonal specifically detects FOXR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FOXR2 | |
Polyclonal | |
Purified | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Forkhead box protein N6, forkhead box protein R2, forkhead box R2, FOXN6MGC21658 | |
FOXR2 | |
IgG | |
Protein A purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
NP_940853 | |
139628 | |
Synthetic peptide directed towards the N terminal of human FOXR2. Peptide sequence VDPNILCPLGSQEAPKPSGKEDLTNISPFPQPPQKDEGSNCSEDKVVESL. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title