Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXRED1 Polyclonal Antibody
FOXRED1 Polyclonal Antibody
Supplier: Thermo Scientific PA565242
Description
FOXRED1 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
FOXRED1 | |
Polyclonal | |
Unconjugated | |
FOXRED1 | |
BC024806; FAD dependent oxidoreductase domain containing 1; FAD-dependent oxidoreductase domain containing 1; FAD-dependent oxidoreductase domain-containing protein 1; Foxred1; FP634; H17; RGD1311785; TEG-23; testis expressed gene 23; Tex23 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55572 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96CU9 | |
FOXRED1 | |
Recombinant Human FOXRED1. Recombinant protein control fragment (Product #RP-97317). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction