Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPR A14, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
329691-12-5
C23H20N2O5
404.422
ULOKADSYVZOTTL-CFRMEGHHSA-N
fpr a14,1,3-benzodioxolane-5-carboxylic acid 4'-benzyloxy-3'-methoxybenzylidene hydrazide,n'-z-4-benzyloxy-3-methoxyphenyl methylidene-1,3-benzodioxole-5-carbohydrazide,n-z-3-methoxy-4-phenylmethoxyphenyl methylideneamino-1,3-benzodioxole-5-carboxamide,n-3-methoxy-4-phenylmethoxy phenyl methylideneamino-1,3-benzodioxole-5-carboxamide,2h-benzo 3,4-d 1,3-dioxolan-5-yl-n-1z-2-3-methoxy-4-phenylmethoxy phenyl-1-azavinyl carboxamide
5503194
N-[(Z)-(3-methoxy-4-phenylmethoxyphenyl)methylideneamino]-1,3-benzodioxole-5-carboxamide
COC1=C(C=CC(=C1)C=NNC(=O)C2=CC3=C(C=C2)OCO3)OCC4=CC=CC=C4
This item is not returnable. View return policy
329691-12-5
C23H20N2O5
404.422
ULOKADSYVZOTTL-CFRMEGHHSA-N
fpr a14,1,3-benzodioxolane-5-carboxylic acid 4'-benzyloxy-3'-methoxybenzylidene hydrazide,n'-z-4-benzyloxy-3-methoxyphenyl methylidene-1,3-benzodioxole-5-carbohydrazide,n-z-3-methoxy-4-phenylmethoxyphenyl methylideneamino-1,3-benzodioxole-5-carboxamide,n-3-methoxy-4-phenylmethoxy phenyl methylideneamino-1,3-benzodioxole-5-carboxamide,2h-benzo 3,4-d 1,3-dioxolan-5-yl-n-1z-2-3-methoxy-4-phenylmethoxy phenyl-1-azavinyl carboxamide
5503194
N-[(Z)-(3-methoxy-4-phenylmethoxyphenyl)methylideneamino]-1,3-benzodioxole-5-carboxamide
COC1=C(C=CC(=C1)C=NNC(=O)C2=CC3=C(C=C2)OCO3)OCC4=CC=CC=C4

FPR agonist

Formyl peptide receptor (FPR) agonist that potently activates neutrophils in vitro (EC 50 values are 42 and 630 nM for neutrophil chemotaxis and Ca 2+ mobilization respectively).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 404.42
Percent Purity >99%
Chemical Name or Material FPR A14

Recommended Storage : Store at Room Temperature