Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPR A14, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
329691-12-5
C23H20N2O5
404.422
ULOKADSYVZOTTL-CFRMEGHHSA-N
fpr a14, 1,3-benzodioxolane-5-carboxylic acid 4'-benzyloxy-3'-methoxybenzylidene hydrazide, n'-z-4-benzyloxy-3-methoxyphenyl methylidene-1,3-benzodioxole-5-carbohydrazide, n-z-3-methoxy-4-phenylmethoxyphenyl methylideneamino-1,3-benzodioxole-5-carboxamide, n-3-methoxy-4-phenylmethoxy phenyl methylideneamino-1,3-benzodioxole-5-carboxamide, 2h-benzo 3,4-d 1,3-dioxolan-5-yl-n-1z-2-3-methoxy-4-phenylmethoxy phenyl-1-azavinyl carboxamide
5503194
N-[(Z)-(3-methoxy-4-phenylmethoxyphenyl)methylideneamino]-1,3-benzodioxole-5-carboxamide
COC1=C(C=CC(=C1)C=NNC(=O)C2=CC3=C(C=C2)OCO3)OCC4=CC=CC=C4
This item is not returnable. View return policy
329691-12-5
C23H20N2O5
404.422
ULOKADSYVZOTTL-CFRMEGHHSA-N
fpr a14, 1,3-benzodioxolane-5-carboxylic acid 4'-benzyloxy-3'-methoxybenzylidene hydrazide, n'-z-4-benzyloxy-3-methoxyphenyl methylidene-1,3-benzodioxole-5-carbohydrazide, n-z-3-methoxy-4-phenylmethoxyphenyl methylideneamino-1,3-benzodioxole-5-carboxamide, n-3-methoxy-4-phenylmethoxy phenyl methylideneamino-1,3-benzodioxole-5-carboxamide, 2h-benzo 3,4-d 1,3-dioxolan-5-yl-n-1z-2-3-methoxy-4-phenylmethoxy phenyl-1-azavinyl carboxamide
5503194
N-[(Z)-(3-methoxy-4-phenylmethoxyphenyl)methylideneamino]-1,3-benzodioxole-5-carboxamide
COC1=C(C=CC(=C1)C=NNC(=O)C2=CC3=C(C=C2)OCO3)OCC4=CC=CC=C4

FPR agonist

Formyl peptide receptor (FPR) agonist that potently activates neutrophils in vitro (EC50 values are 42 and 630 nM for neutrophil chemotaxis and Ca2+ mobilization respectively).
Quantity 50 mg
Formula Weight 404.42
Percent Purity >99%
Chemical Name or Material FPR A14

Recommended Storage : Store at Room Temperature