Learn More
FPRL1/FPR2 Mouse anti-Human, Alexa Fluor 350, Clone: 304405, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB3479U100UG
Catalog No. FAB3479U100
Add to cart
This item is not returnable. View return policy
Description
Description
FPRL1/FPR2 Monoclonal specifically detects FPRL1/FPR2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
FPRL1/FPR2
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P25090
FPR2
NS0 mouse myeloma cell line transfected with human FPRL1 Met1-Met351 Accession # P25090
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains FPRL1 transfectants but not irrelevant transfectants. It also stains monocytes and granulocytes.
Flow Cytometry
304405
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
ALXR, FMLP-R-I, FMLP-R-II, FMLPX, formyl peptide receptor 2, Formyl peptide receptor-like 1RFP, FPR2, FPR2A, FPRH1FMLP-related receptor I, FPRL1LXA4 receptor, HM63, HM63FPRH2, Lipoxin A4 receptor, lipoxin A4 receptor (formyl peptide receptor related), LXA4 Receptor, LXA4RFMLP-R-I, N-formyl peptide receptor 2, RFP
Mouse
IgG2b
RUO
Primary
2358
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)