Learn More
Canon Instrument Co FPRM FLASHPOINT REF 10 155G
SDP
Manufacturer:  Canon Instrument Co 9727A10
Encompass_Preferred
CANNON flash point reference materials are produced in accordance with ASTM D56, Tag Closed Cup Flash Point, ASTM D92, Cleveland Open Cup Flash Point and ASTM D93, Pensky Marten Closed Cup Flash Point. Lot specific data including average flash point and observed standard deviation from a statistical analyis of 40 tests in 20 different laboratories is provided with each standard. Flash Point Reference Materials are sold in 200 mL bottles. *HAZARD* FLASH POINT FPRM10 n-DECANE 200ml (155g)
Catalog No. NC9819994