Learn More
FPRP/PTGFRN Mouse anti-Human, Alexa Fluor 750, Clone: 998107, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB100431S100UG
Catalog No. FB10431S100
Add to cart
This item is not returnable. View return policy
Description
Description
FPRP/PTGFRN Monoclonal antibody specifically detects FPRP/PTGFRN in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
FPRP/PTGFRN
998107
Flow Cytometry 0.25-1 μg/106 cells
Q9P2B2
CD315, CD9P-1, FPRP, PTGFRN
Human embryonic kidney cell, HEK293-derived human FPRP, Pro27-Pro832, Accession # Q9P2B2
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human FPRP in direct ELISAs.
Flow Cytometry
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG1
100 μg
Primary
5738
Human