Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ FR Control 2.0™ Tan Short Sleeve T-Shirt
SDP

Catalog Number p-7163234
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

National Safety Apparel™ FR Control 2.0™ T-Shirt is designed to proactively regulate skin temperature.

  • CAT 1
  • Fabric/Material: 5.5 oz. Modacrylic Blend
  • Proactively regulates skin temperature (± 3 degrees)
  • Reduces perspiration by as much as 34%
  • Stretch and flexibility to maintain a comfortable work environment
  • Heat stress reduction
  • Odor control
  • Tagless shirt collar
  • Superior moisture wicking and dry rate
  • Inherently flame resistant
  • FR protection will not wash/wear out
  • Lightweight, breathable fabric
  • Knit wrists keep sleeves in place
  • Made in the USA
Size Large
Material Modacrylic Blend
Color Tan
Sleeve Style Short Sleeve
Arc Flash Rating 4 cal/cm2
Certifications/Compliance NFPA 70E for Arc Flash, ASTM F1506 for Arc Flash, CSA Z462 for Arc Flash (Canada), OSHA 1910.269
For Use With (Application) FR base layer
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit